missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD Hybridoma AGT™ Medium
Description
Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.
- Ability to support suspension and stationary hybridomas and myelomas
- Formulated without L-glutamine for stability
- Animal origin-free, protein-free and chemically defined formulation
- Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Specifications
Specifications
| Product Type | CD Hybridoma Medium |
| Concentration | 1 X |
| Additive | Sodium Pyruvate |
| Buffer | Sodium Bicarbonate, No HEPES |
| Cell Type | Myeloma Cells, Hybridomas |
| Content And Storage | Storage conditions: 2-8°C, dark and dry Shipping conditions: Gel packs Shelf life: 24 months from date of manufacture |
| Classification | Animal Origin-Free, Chemically Defined, Protein-Free |
| Culture Environment | CO2 |
| Endotoxin Level | Low |
| Glucose Concentration | 3.66 g/L |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?