Learn More
Gibco™ TNF Recombinant Human Protein
Product Code. 10630301
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Product Code. 10630301
missing translation for 'mfr': Gibco™ CTP3013
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Requires
Recombinant TNF is a bioactive protein intended for use in cell culture applications. TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.
TNF; Recombinant Human Protein
  • TNF causes cytolysis and cytostasis of many tumor cell lines
  • With wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis
  • ED50 range = 0.02 to 0.05ng/mL (Specific Activity: 2 x 107 to 5 x 107 units/mg), determined by dose dependent cytotoxic effect on L929 cells in presence of actinomycin D
Cell Culture, Dendritic Cell Culture, Hematopoietic Stem Cell Culture, Mammalian Cell Culture, Mesenchymal Stem Cell Culture, Primary Cell Culture, Stem Cell Culture, Stem Cell Research
Order Info
Shipping Condition: Wet ice
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
For Research Use Only. Not for use in diagnostic procedures.