missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Flt-3 Ligand (FLT3L) Recombinant Protein
Fms-Like Tyrosine Kinase 3 Ligand (FLT3 Ligand, FLT3LG) is a bioactive protein intended for use in cell culture applications
428.00€ - 6510.00€
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 35 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | FLT-3 Ligand |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10162288
|
Gibco™
PHC9414 |
10μg |
428.00€
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10631120
|
Gibco™
PHC9415 |
25μg |
762.00€
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10365588
|
Gibco™
PHC9411 |
100μg |
1576.00€
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10688118
|
Gibco™
PHC9413 |
1mg |
6510.00€
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human Flt-3 Ligand (FLT3L) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
FLT3 Ligand is a cytokine involved in hematopoeisis.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human OCI-AML5 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Human | |
Mammalian | |
Recombinant, Ligand | |
FLT-3 Ligand | |
Gibco™ |
35 kDa | |
FLT-3 Ligand | |
Lyophilized Human Flt-3 Ligand should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Flt-3 Ligand at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum. | |
< 0.1 ng/μg | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Signal Transduction, Stem Cell Research, Oncology | |
>95% by SDS-PAGE, >95% by SDS-PAGE |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.