missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ GM-CSF Recombinant Rat Protein

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Marque:  Gibco™ PRC2015

Informations supplémentaires : Poids : 0.50000kg

 Afficher plus de versions de ce produit

Code produit. 10009182

  • 222.00€ / 1 pièce


Date dexpédition estimée
du fournisseur 13-02-2020
Ajouter au panier

Description

Description

Recombinant rat GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Shipping Condition: Wet Ice.

Spécification

Spécification

Cell Culture; Mammalian Cell Culture
116630
Exchange Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
14.6kDa
95%
Rat
Carrier Protein
GM-CSF, G-CSF, & M-CSF
GM-CSF
Cytokines & Receptors
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.