Learn More
Gibco™ IGFBP6 Recombinant Human Protein
Recombinant Human Insulin-like Growth Factor Binding Protein 6 (IGFBP6) is a bioactive protein intended for use in cell culture applications
POA
Specifications
Gene Alias IGFBP-6
View More Specs
Description
Description
IGFBP6 binds IGF-1 and II, prolonging the half life of these growth factors and altering the way these growth factors interact with cells surface receptors.
  • High purity: Minimizes interference from other proteins or contaminants
  • High biological activity: More results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: To help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
  • High Biological Activity: In-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent inhibition of IGF-II induced proliferation of MCF-7 cells
Shipping Condition: Wet Ice.
Specifications
Specifications
IGFBP-6
Documents
Product Certifications
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.