Learn More
Gibco™ Rat CCL5 (RANTES) Recombinant Protein
Rat CCL5 (RANTES) Recombinant Protein
Brand: Gibco™ PRC1054
Additional Details : Weight : 0.50000kg
Description
Rat CCL5 (RANTES) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
7.9 kDa | |
RANTES | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Chemokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Oncology | |
98 % |
81780 | |
20 ug | |
Rat | |
Carrier-Free | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
Rantes | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.