Learn More
Gilson™ MICROMANTM E Capillary Pistons
GREENER_CHOICE
Product Code. 10463942 Shop All Gilson Products
Click to view available options
Quantity:
10 x 96 Pk.
182 Pk.
192 Pk.
200 Pk.
576 Pk.
960 Pk.
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
CP10
CP100
CP1000
CP1000ST
CP100ST
CP10ST
CP25
CP250
CP250ST
CP25ST
CP50
CP50ST
Format:
Box
Rack, 10 x 96
Rack, 2 x 96
Unit Size:
Pack of 182
Pack of 192
Pack of 200
Pack of 576
Pack of 960
Product Code. 10463942
Brand: Gilson™ F148112
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
MICROMANTM E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for GilsonTM MICROMANTM E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 3 to 25 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model CP25
Length (English) 4.25 in.
Length (Metric) 86.2 mm
Format Box
Tip Style Positive Displacement
Packaging Box
Quantity 200 Pk.
Product Type Capillary Piston
Disposable Single-use
For Use With (Equipment) Gilson Microman
Material Polypropylene
Fit for Purpose Certified Quality