Learn More
Gilson™ PIPETMAN Neo™ Multichannel Pipets
Shop All Gilson Products
Click to view available options
Volume (Metric):
2 to 20 μL
20 to 200 μL
Number of Channels:
8
12
Model:
P12x200N
P12x20N
P8x200N
P8x20N
Unit Size:
Each
Description
- Easy-to-remove tip ejector with autoclavable lower part
- Internal design applies equal pressure on each piston to ensure consistent volume levels across all channels
- PVDF handle minimizes the hand-warming effect during pipetting and is resistant to aggressive chemicals
- Gilson Trident and unique spacer create a stepped ejection, staggering the tip ejection and thus reducing the force required to eject tips
- Unique tip holder allows for a secure fit of most brands of tips
- Available in 8- and 12-channel models
Order Info
Trade-in program available. Please contact your Fisher Scientific Sales representative for more details.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Model | P8x20N |
| Number of Channels | 8 |
| Product Line | Pipetman Neo |
| Type | Multichannel Pipette |
| Volume (Metric) | 2 to 20 μL |
| Warranty | 1 Year |