Learn More
Liebherr™ GKv 5710 Model GN 2/1 Refrigerator
Product Code. 10452468 Shop All Liebherr Products
Click to view available options
Capacity (Metric):
583 L
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 10452468
Brand: Liebherr™ GKV 5710
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Specifications
Type General Purpose Refrigerator
Capacity (Metric) 583 L
Temperature Range 1°C to 15°C
Refrigerant R 600a
Door Style SwingLine
Hinge Side Right
Shelves 6
Capacity (Metric) Shelf 60 kg
Cabinet Material Plastic
Display Digital
Color White
Control Type Electronic
Includes 6 Shelves
Defrost Automatic
Lid Yes
Model GKv 5710
Dimensions (DxWxH) Exterior 186.4 H x 75 W x 75 Dcm
Dimensions (D x W x H) Interior 58.1 x 65 x 151 cm
Depth (Metric) Exterior 75 cm
Depth (Metric) Interior 58.1 cm
Height (Metric) Exterior 186.4 cm
Height (Metric) Interior 151 cm
Width (Metric) Exterior 75 cm
Width (Metric) Interior 65 cm
Voltage 220/240 V
Electrical Requirements 220 - 240 V 50 Hz 1 A 100 w
Frequency 50 Hz
Wattage 100 w
Energy Usage 2.152kWh/24 hr.
Refrigeration System Voltage 220 to 240V
Shipping Weight (Metric) 96 kg
Weight (Metric) 89 kg