Learn More
BRAND™ Glass Automatic Burette - BB
Product Code. 15805522 Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Product Code. 15805522
Brand: BRAND™ 922523
Only null left
Add to basket
Add to basket
Without intermediate stopcock. Includes bottle.
  • Safety stopcock
  • Individual conformity certificate
  • TRUSTED_SUSTAINABILITY
    Specifications
    Capacity (Metric) 50 mL
    Material Glass
    Graduations 0.1 mL
    Includes Bottle
    Type Automatic Buret