Learn More
Hecht Karl™ Glass Balls
Product Code. 10699421
Click to view available options
Diameter (Metric):
1 mm
2 mm
4 mm
6 mm
Unit Size:
1kg
Product Code. 10699421
Brand: Hecht Karl™ 41401006
Only null left
Add to basket
Edge
Add to basket
Made of clear, polished glass. Karl Hecht™ Glass Balls can be used as boiling stones, mixing beads or for distillation columns.
 Spherical, solid, polished glass beads for general laboratory use.
Specifications
Type Ball
Material Glass
Diameter (Metric) 6 mm
Quantity 1kg Pack