Learn More
Hecht Karl™ Glass Class AS Volumetric Pipet
Brand: Hecht Karl™ 42556052
Additional Details : Weight : 0.25000kg
Specifications
Yes | |
AS | |
Yes | |
1 Pack | |
10 mL |
DIN EN ISO 648, Conformity certified | |
Glass | |
Amber | |
0.02 mL | |
Volumetric Pipet |