Learn More
Sonics Materials™ Glass Cooling Cell
222.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10223302
|
Sonics Materials™
830-00001 |
300 mL |
222.00€
Each |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10305222
|
Sonics Materials™
830-00003 |
30 mL |
222.00€
Each |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Basic Disperser | |
1 Ea. |