Learn More
Rogo Sampaic™ Glass Coplin Staining Jar
Product Code. 11702654
Click to view available options
Slide Count:
5
Unit Size:
Each
Product Code. 11702654
Brand: Rogo Sampaic™ BPL005
Only null left
Add to basket
Edge
Add to basket
Holds 10 microscope slides of approx. 76 x 26mm. Rogo Sampaic™ Glass Coplin Staining Jars are manufactured from soda lime glass.
  • Clear glass
  • High wall shape
  • With screw cap
Specifications
Material Glass
Type Coplin Staining Jar
Slide Count 5
Dimensions (L x W) 45 x 45 mm
Length (Metric) 45 mm
Width (Metric) 45 mm
Depth (Metric) 105 mm