Learn More
Hirschmann™ Glass Coverslips for Haemacytometer
Product Code. 10099780
Click to view available options
Dimensions (L x W):
20 x 26 mm
Product Type:
Hemacytometer Coverslip
Unit Size:
Pack of 10
Product Code. 10099780
Brand: Hirschmann™ 8000124
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Cover glasses for Haemacytometer with smooth surface grinding, without CE mark.
  • Surface ground and polished surface
  • LOT number on the packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC According to the German Calibration Regulation (Eichordnung), only officially calibrated haemocytometer glass covers may be placed on the market and used in the FRG
Specifications
Product Type Hemacytometer Coverslip
Material Glass
Thickness (Metric) 0.4 mm
Dimensions (L x W) 20 x 26 mm
For Use With (Equipment) Haemocytometer