Learn More
Fisherbrand™ Glass-Encased Stir Bar
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
6 mm
12 mm
19 mm
25 mm
45 mm
60 mm
Diameter (Metric):
5 mm
6 mm
8 mm
Unit Size:
Pack of 10
Pack of 5
Description
- Stable, general-purpose magnetic stir bar
- Use with very abrasive media that may erode PTFE
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | Clear |
| Description | Glass encapsulated Alnico V magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 25 mm |
| Quantity | 5/Pk. |
| Shape | Cylinder |