Learn More
Fisherbrand™ Glass-Encased Stir Bar
Product Code. 11878922 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
6 mm
12 mm
19 mm
25 mm
45 mm
60 mm
Diameter (Metric):
5 mm
6 mm
8 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 11878922
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use with very abrasive media that may erode PTFE. The glass-encased magnetic stir bar is cylindrical for a wide variety of uses.
  • Stable, general-purpose magnetic stir bar
  • Use with very abrasive media that may erode PTFE
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Clear
Description Glass encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 5 mm
Length (Metric) 6 mm
Quantity 5/Pk.
Shape Cylinder