Learn More
Merck Glass Fiber Filters Without Binder
Product Code. p-108666 Shop All Merck Products
Change view
Click to view available options
Diameter (Metric):
1 cm
2.4 cm
2.5 cm
3.7 cm
4.7 cm
7 cm
9 cm
124 mm
14.2 cm
20.3 cm
Grade:
AP40
APFF
Quantity:
100 Pk.
50 Pk.
500 Pk.
Unit Size:
Pack of 100
Pack of 50
Pack of 500
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Grade Quantity unitSize
10635591 3.7 cm AP40 500 Pk. Pack of 500
10598442 2.4 cm AP40 500 Pk. Pack of 500
10379980 14.2 cm AP40 50 Pk. Pack of 50
10498603 2.5 cm AP40 100 Pk. Pack of 100
10004570 4.7 cm AP40 500 Pk. Pack of 500
10572844 4.7 cm AP40 100 Pk. Pack of 100
10588442 1 cm AP40 100 Pk. Pack of 100
10437221 14.2 cm APFF 50 Pk. Pack of 50
10697121 2.5 cm APFF 100 Pk. Pack of 100
11758792 124 mm APFF 50 Pk. Pack of 50
10480021 9 cm APFF 50 Pk. Pack of 50
10340131 9 cm AP40 100 Pk. Pack of 100
10498793 7 cm AP40 100 Pk. Pack of 100
10257770 4.7 cm APFF 100 Pk. Pack of 100
11768772 20.3 cm AP40 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 10635591 Supplier Merck Supplier No. AP4003705
Only null left
Add to basket
Add to basket
Choose from six grades of filters. Glass Fiber Filters without Binders are made of borosilicate microfiber glass. Available in multiple pore sizes and flow rates for a variety of filtration and analysis applications utilizing depth filters. Autoclavable.
For prefiltration and contamination analysis
OverviewMillipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).Glass Fiber Filters with Binder Resin:Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.
Type AP15
  • Lowest dirt-holding capacity
  • Recommended prefilter for 0.2 to 0.6 μm filters
Type AP20
  • Lower retention, higher dirt-holding capacity than AP15 filters
  • Recommended prefilter for 0.8 to 8.0 μm filters
  • Use upstream to protect type AP15 filters
Type AP25
  • Added thickness yields same retention and greater dirt-holding capacity as AP20
  • Recommended prefilter for 0.9 to 8 μm filters, especially for proteinaceous and heavily contaminated liquids
  • Use upstream to protect type AP15 filters Glass Fiber Filters without Binder Resin:The filters without binder resin retain their structural integrity without weight loss when heated up to 500°C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
Type APFA
  • Retains fine particles with good efficiency, even at high flow rates
  • Recommended for monitoring wastewater and collecting suspended particles in gases (e.g., fumes from chimneys)
  • Use in biochemical applications (e.g., collection of cells and filtration of protein or nucleic acid precipitates)
Type APFB
  • Higher mechanical strength when wet and greater loading capacity than type APFA filters
  • Use for liquid clarification, quantification of solids in suspensions of fine particles, and scintillation counting
Type APFC
  • Greater retention, especially for fine particulate and microorganism removal than type APFA filters
  • Use for determining total suspended solids in drinking water
  • Use for filtering proteins or nucleic acid TCA precipitates and for collecting cells and microorganisms
Type APFD
  • Thick filter with high flow and low retention
  • Use for clarifying suspensions containing particulates greater than 1.0 μm
Type APFF
  • Use for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates
  • Recommended for USEPA method 1311 for TCLP analysis
Type AP40
  • Recommended for Standard Methods Total Suspended Solids 2540D
  • Recommended for USEPA method 1311 for TCLP analysis
  • Maintains structural integrity without weight loss when ignited to 550°C (1022°F) after sample filtration
  • Recommended for determining volatile suspended matter in wastewater and industrial effluents
TRUSTED_SUSTAINABILITY
Specifications
Airflow Rate 44.5 L/min/ cm2
Air Resistance 50 mm of H2O
Autoclavable Autoclavable
Color White
Filter Type Depth Filter
Material Borosilicate Glass Microfiber
Pore Size 0.7 μm
Sterility Sterile
Flow Rate 6 mL/min/ cm2
For Use With (Application) Determination of volatile suspended matter in wastewater and industrial effluents
Grade AP40
Hydrophobicity Hydrophilic
Quantity 500 Pk.
Surface Type Plain
Temperature (English) Max. 1022°F
Temperature (Metric) Max 550°C
Thickness (English) 0.018 in.
Thickness (Metric) 475 μm
Type Filter Membranes
Wettability Hydrophilic
Diameter (English) 1.4 in.
Diameter (Metric) 3.7 cm
Diameter (Metric) Disc 3.7 cm