missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glass Fiber GF8 Filter
Description
This glass fibre filter has an inorganic binder for the filtration of coarse particles. Frequently used in environmental analysis for the determination of PCB, DDE, DDT, furans, and dioxins in the air; pollution measurements in industrial, urban and populated areas, cement factories, the iron and steel industry; dust measurements at the workplace; determination of the dust fraction in technical gases; and testing the effectiveness of dust-collecting.
Specifications
Specifications
| Airflow Rate | 12 s/100 mL/1.56 cm2 |
| Description | GE’s Whatman Grade GF 8 glass microfiber filters are used for filtration of coarse particles. GF 8 offers faster filtration and a higher air flow rate compared to the similar Grade GF 9 filters. |
| Format | Disc |
| Material | Borosilicate Glass |
| Filtration Time | Fast |
| For Use With (Application) | Used in the filtration of coarse particles.Frequently used in environmental analysis, in the determination of PCB, DDE, DDT, furans and dioxins in the air; pollution measurements in industrial, urban and populated areas, cement factories, iron and steel |
| Grade | Grade GF 8 |
| Quantity | 100 Pack |
| Type | GF8 Filter |
| Diameter (Metric) | 200 mm |
Safety and Handling
Product Identifier
- FILTER GLASS CIRCLE
- Warning
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?