Learn More
Rogo Sampaic™ Glass Laveran Staining Jar
Product Code. 11722654
Click to view available options
Slide Count:
3
Unit Size:
Each
Product Code. 11722654
Brand: Rogo Sampaic™ BPL031
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Material Glass
Type Staining Jar
Slide Count 3
Dimensions (L x W) 58 x 94 mm
Length (Metric) 58 mm
Width (Metric) 94 mm
Depth (Metric) 33 mm