missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Glass Microfiber Filters, Discs, Grade 13440

Binder-free glass microfiber filters for analytical and gravimetric analyses and prefiltering.

Brand:  Sartorius Stedim Biotech™ 13440-100------K

Additional Details : Weight : 0.13500kg

Product Code. 15942480

  • 116.00€ / Pack of 50
Estimated Shipment: 29-08-2024
to see stock.
Add to basket

Description

Description

  • Binder-free
  • Suitable for the following:
    • Prefiltration of relatively clean solutions such as tissue culture media.
    • Analysis of suspended solids in wastewater according to EN 872.
    • Analytical sample preparation
    • Particle analysis
    • Clarification
  • Non-sterile, bulk packed
Specifications

Specifications

88 g/m2
Binder-free Glass Microfiber
Hydrophilic
500°C
Disc
13400
50/Pk.
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions