Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.
Brand:  Hirschmann™ 9000240
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10480771
  • 132.00€ / Pack of 1000
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • End-to-end, with minicaps
  • Conformity certified
  • Available either heparinized or non-heparinized
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
0.5%
DURAN
30 ±0.5 mm
Single-use
ISO 7550, CE, IVD 98/79 EC
40 μL
Minicap Capillary Pipette