Learn More
Hirschmann™ Glass Stopper
Product Code. 30090403
Click to view available options
Color:
Clear
Unit Size:
Pack of 10
Product Code. 30090403
Brand: Hirschmann™ 8900370
Only null left
Add to basket
Add to basket
Hirschmann™ Glass Stopper is excellent for tight sealing of standard ground vessels.
  • For tight sealing of standard ground vessels.
    .
Specifications
Standard Taper Joint NS 14/23
Color Clear
Material Glass