Learn More
DWK Life Sciences Glass Stopper, Clear, SBW Glass, Short Standard Taper Ground Joint, Octagonal flat head
Clear SBW glass ground stopper, short-length
Brand:  DWK Life Sciences 246250605
Additional Details : Weight : 0.17000kg
 View more versions of this product
Product Code. 15241989
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Solid or semi-hollow, short-length, clear SBW glass stoppers can be used to close glassware items such as flasks or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head.
  • All glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Octagonal shaped head
Specifications
Specifications
Solid
10/Pk.
ST 14/15
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
SBW Glass
No