Learn More
Biochrom Glass Test Tubes
Glass Test Tubes for use with Ultrospec 7000/8000/9000 and Novaspec III/Plus. Require additional accessories for use.
Brand:  Biochrom 80-2004-50
Additional Details : Weight : 0.01000kg
Product Code. 11843194
  • 211.00€ / Pack of 10
Estimated Shipment: 22-10-2024
to see stock.
Add to basket
Description
Description
  • Glass test tubes
  • 12mm dia. x100mmL
  • For use with Ultrospec 7000/8000/9000
    • Requires Tube Holder 80-7100-11
  • For use with Novaspec III/Plus
    • Requires Tet Tube Adapter 80-2117-47
Specifications
Specifications
Test Tube
Glass
10 Pk.