Learn More
BUCHI Glass Vapor Ducts with Combi-Clip
Replacement vapor duct tubes for BUCHI rotary evaporators.
Brand: BUCHI 032339
Additional Details : Weight : 0.11700kg
Description
- Fabricated from borosilicate glass
- ST joint number 29/32
Specifications
Glass | |
Vapor Duct with Combi-Clip | |
SJ29/32 | |
1 Each |