Learn More
Duran™ Glass Volumetric Flask
With beaded rim
Brand: Duran™ 213728673
141.17 EUR valid until 2024-05-31
Use promo code "21660" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Without stopper bed
- No print
Specifications
10 L | |
Glass | |
with beaded rim |