Learn More
Rogo Sampaic™ Class B Glass Volumetric Pipettes
Click to view available options
Volume (Metric):
2 mL
10 mL
20 mL
25 mL
50 mL
Color:
Black
Orange
Red
Yellow
Unit Size:
Each
Pack of 2
Description
- With vitrified amber graduations
- With safety bulb
Specifications
Specifications
| Accuracy | 0.02 mL |
| Calibrated | Yes |
| Calibration | 0.01mL |
| Class | B |
| Graduated | Yes |
| Graduation Color | Brown |
| Graduations | 0.01 mL |
| Color | Black |
| Volume (Metric) | 2 mL |
| Length (Metric) | 360 mm |