Learn More
Hofmann Klaus™ Glass Weighing Boats With Stem
Product Code. 11920787
Click to view available options
Capacity (Metric):
6 mL
Unit Size:
Each
Product Code. 11920787
Brand: Hofmann Klaus™ 24301.07
Only null left
Specifications
Bottom Shape Flat
Capacity (Metric) 6 mL
Material Glass
Shape Boat
Type Weighing Boat
For Use With (Application) Ideal for lossless pouring of the sample
Size 2
Length (Metric) 70 mm