Learn More
Glassy carbon rod, 1mm (0.04in) dia, type 1, Thermo Scientific Chemicals
Widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.
Brand: Thermo Scientific Alfa Aesar 038009.DR
Additional Details : Weight : 0.01000kg
Description
Glassy carbon rod is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
MFCD00133992 | |
Insoluble in water. |
Tube | |
200 mm |
RUO – Research Use Only