missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon rod, 3mm (0.1in) dia, type 1, Thermo Scientific Chemicals

Catalog Number 15421837
Click to view available options
Quantity:
50 mm
100 mm
200 mm
7440-44-0
MFCD00133992
7440-44-0
MFCD00133992

Used as an electrode material in electrochemistry. It is also employed as an electrode material for the fabrication of sensors. It serves as an ideal material for the vacuum technology. It is also used in casting, smelting of metal, hard alloys and high temperature crucibles.

Glassy carbon rod is used as an electrode material in electrochemistry. It is also employed as an electrode material for the fabrication of sensors. It serves as an ideal material for the vacuum technology. It is also used in casting, smelting of metal, hard alloys and high temperature crucibles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Tube
Quantity 200 mm
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon rod

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'