Learn More
Glassy carbon rod, 7mm (0.28 in.) dia, type 2, Thermo Scientific Chemicals
Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.
Brand: Thermo Scientific Alfa Aesar 037999.DR
Additional Details : CAS : 7440-44-0 Weight : 0.01000kg
Description
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
Tube | |
200 mm |
7440-44-0 | |
MFCD00133992 | |
Insoluble in water. |
RUO – Research Use Only