missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon rod, 7mm (0.28 in.) dia, type 2, Thermo Scientific Chemicals

Catalog Number 11374307
Click to view available options
Quantity:
50 mm
100 mm
200 mm
This item is not returnable. View return policy
7440-44-0
MFCD00133992
This item is not returnable. View return policy
7440-44-0
MFCD00133992

Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.

Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Tube
Quantity 50 mm
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon rod

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'