missing translation for 'onlineSavingsMsg'
Learn More

Glauberite, naturally occuring mineral, pieces, ≈1 in., Thermo Scientific™

Catalog Number 15496595
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
13767-89-0
CaH4Na2O8S2
282.20
MFCD05663779
GYZGFUUDAQXRBT-UHFFFAOYSA-N
calcium disodium bis sulfuric acid
This item is not returnable. View return policy
13767-89-0
CaH4Na2O8S2
282.20
MFCD05663779
GYZGFUUDAQXRBT-UHFFFAOYSA-N
calcium disodium bis sulfuric acid

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Packaging Plastic bottle
Quantity 1 Pc.
Formula Weight 278.18
Chemical Name or Material Glauberite
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'