missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes, Purple

Conform well to irregular, curved, rough and highly textured surfaces. Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes are perfect for harsh, industrial environments.

697.00€

Specifications

Description Suggested Ribbon Series: R10000
Adhesive Type Acrylic, Permanent, Cold-temperature
Coating None
Color Purple
Finish Gloss
View More Specs

Products 1
Product Code Brand Width (English) Tape Price Quantity & Availability  
Product Code Brand Width (English) Tape Price Quantity & Availability  
15252936
View Documents
Brady™
113147
2.25 in.
697.00€
Pack of 1
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2026
Add to basket
 
Description

Description

  • Adhere to difficult low surface energy items
  • 5 year outdoor durability
  • Sticks to high textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes
Specifications

Specifications

Suggested Ribbon Series: R10000
None
Gloss
Vinyl Film (B-595)
Non-sterile
GlobalMark™ Label Maker
-40°C
-40°F to +180°F
0.004 in.
100 ft.
Acrylic, Permanent, Cold-temperature
Purple
Continuous
Thermal Transfer
Arc Flash Labels, Arrow Tape, Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking
1
1 Roll/Box
-40°C to +82°C
0.102 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.