Learn More
Plas Labs™ Glove Box 855-ACB Anaerobic Chamber
Product Code. 10550293
Click to view available options
Voltage:
120 V
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10550293
Brand: Plas Labs™ 855ACB/EXP
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Manually control your atmosphere when working with oxygen-sensitive materials in this glove box. Plas Labs™ Glove Box 855-ACB Anaerobic Chamber uses a catalyst heater to reduce any trace entering oxygen to water vapor.
  • Standard 24-month warranty (not applicable for gloves and consumables)
  • Formed one-piece clear acrylic top
  • Self-sealing quick disconnects enable the operator to change the drying train without disturbing the internal atmosphere
  • Matched die-molded thermoset white bottom
  • Adjustable vacuum gauge on transfer chamber
  • Two vacuum pressure pumps, one each for transfer chamber and drying train
  • Four ground key cock valves for purging
  • Drying train includes polycarbonate canisters containing DesiSphere™
  • Inner door on transfer chamber opens automatically when pressures are neutralized between main chamber and transfer chamber
  • All controls are illuminated
  • Transfer chamber is 12in. long and 11in. in diameter (I.D.)
  • Bright Light system (40,000 hour lamp guarantee)
  • Electrical outlet strip; U.L. & C.S.A. approved, hospital grade with four outlets
  • White ambidextrous Hypalon gloves
  • All clamps are adjustable to compensate for wear
Specifications
Depth (Metric) 119 cm
Depth (Metric) Interior 97 cm
Height (Metric) 107 cm
Height (Metric) Interior 79 cm
Hertz 60 Hz
Voltage 120 V
Volume (Metric) 1157 L
Weight (Metric) 135 kg
Width (Metric) 193 cm
Width (Metric) Interior 152 cm