missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon Like Peptide-2 (1-33), Human

Product Code. 15446899
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
15446899 1 mg 1mg
1 options
This item is not returnable. View return policy

Product Code. 15446899

Brand: Thermo Scientific Chemicals J65432.M^

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 3766.1
Product Type Glucagon Like Peptide-2 (1-33), human
Quantity 1 mg
Regulatory Status RUO - research use only

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.