missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose monohydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.50kg
Pack of 6
Description
- Wide selection of support materials and multiple cartridge/column sizes
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- B&J’s reliable quality
Specifications
Specifications
| Packaging | Plastic Bottle |
| Quantity | 1 kg |
| Residual Solvents | Complying |
| Percent Purity | 97.5 to 102.0% |
| Grade | Reagent Pharmacopeia Organics |
| Physical Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction