Learn More
Glufosinate ammonium, 95%, Thermo Scientific™
Glufosinate ammonium, 95%, CAS # 77182-82-2, also known as glufosinate, is a non-selective phosphorous herbicide.
102.00€ - 324.99€
Chemical Identifiers
| CAS | 77182-82-2 |
|---|---|
| Molecular Formula | C5H15N2O4P |
| Molecular Weight (g/mol) | 198.16 |
| MDL Number | MFCD00055562,MFCD00211362 |
| InChI Key | ZBMRKNMTMPPMMK-UHFFFAOYNA-N |
| Synonym | DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt |
| PubChem CID | 122130249 |
| IUPAC Name | ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate |
| SMILES | [NH4+].CP(O)(=O)CCC(N)C([O-])=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15417449
|
Thermo Scientific Alfa Aesar
J66186.MD |
250mg |
104.00€ 102.00€ / 250mg Save 2.00€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15407449
|
Thermo Scientific Alfa Aesar
J66186.03 |
1g |
332.00€ 324.99€ / 1g Save 7.01€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Glufosinate ammonium is a compound which exhibits broad-spectrum herbicidal properties through inhibition of glutamine synthetase, which disrupts cell membranes
Application
• Glufosinate ammonium acts as a non-selective herbicide
Chemical Identifiers
| 77182-82-2 | |
| 198.16 | |
| ZBMRKNMTMPPMMK-UHFFFAOYNA-N | |
| 122130249 | |
| [NH4+].CP(O)(=O)CCC(N)C([O-])=O |
| C5H15N2O4P | |
| MFCD00055562,MFCD00211362 | |
| DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt | |
| ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate |
Specifications
| 77182-82-2 | |
| >110°C (230°F) | |
| MFCD00055562,MFCD00211362 | |
| 519°C | |
| DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt | |
| ZBMRKNMTMPPMMK-UHFFFAOYNA-N | |
| ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate | |
| 122130249 | |
| 95% |
| White | |
| C5H15N2O4P | |
| 8163399 | |
| 14,7339 | |
| Soluble in water | |
| [NH4+].CP(O)(=O)CCC(N)C([O-])=O | |
| 198.16 | |
| 198.16 | |
| Glufosinate ammonium |
Safety and Handling
GHS H Statement
H360-H373-H302-H312-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H360-H373
missing translation for 'einecsNumber' : 278-636-5
missing translation for 'rtecsNumber' : EK7713600
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.