missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Glycerol
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Description
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis.
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.
Specifications
Specifications
| CAS | 56-81-5 |
| Molecular Formula | C3H8O3 |
| Quantity | 500 mL |
| Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction