missing translation for 'onlineSavingsMsg'
Learn More

Melford Glycerol

Product Code. p-200087161
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 30018280

Brand: Melford G220200.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in sample preparation and gel formation for polyacrylamide gel electrophoresis.

Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.

Specifications

CAS 56-81-5
Molecular Formula C3H8O3
Quantity 500 mL
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.