missing translation for 'onlineSavingsMsg'
Learn More

Glycitein, TRC

Product Code. 30397756
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30397756

Brand: Toronto Research Chemicals TRCG63540010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycitein
CAS 40957-83-3
Purity Grade Notes HPLC
Molecular Formula C16 H12 O5
Quantity 10 mg
Synonym 7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone, Glycetein, Glycitein, Glycitin aglycon
InChI Formula InChI=1S/C16H12O5/c1-20-15-6-11-14(7-13(15)18)21-8-12(16(11)19)9-2-4-10(17)5-3-9/h2-8,17-18H,1H3
SMILES COc1cc2C(=O)C(=COc2cc1O)c3ccc(O)cc3
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-6-methoxychromen-4-one
Molecular Weight (g/mol) 284.26
Formula Weight 284.0685
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.