missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycitein, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Glycitein |
| CAS | 40957-83-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H12 O5 |
| Quantity | 25 mg |
| Synonym | 7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone, Glycetein, Glycitein, Glycitin aglycon |
| InChI Formula | InChI=1S/C16H12O5/c1-20-15-6-11-14(7-13(15)18)21-8-12(16(11)19)9-2-4-10(17)5-3-9/h2-8,17-18H,1H3 |
| SMILES | COc1cc2C(=O)C(=COc2cc1O)c3ccc(O)cc3 |
| IUPAC Name | 7-hydroxy-3-(4-hydroxyphenyl)-6-methoxychromen-4-one |
| Molecular Weight (g/mol) | 284.26 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction