missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-Glutamic Acid, Fisher BioReagents

Product Code. 10695195
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Amber Glass
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 10695195

Brand: Fisher Bioreagents BP2522500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Glycyl-L-Glutamic Acid is used in biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7412-78-4
Molecular Formula C7H12N2O5
Molecular Weight (g/mol) 204.182
InChI Key IEFJWDNGDZAYNZ-BYPYZUCNSA-N
Synonym glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh
PubChem CID 99278
ChEBI CHEBI:73801
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]pentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)NC(=O)CN

Specifications

Color White
Packaging Amber Glass
Quantity 500 mg
Formula Weight 204.18
Physical Form Fluffy Powder
Chemical Name or Material Glycyl-L-Glutamic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.