Learn More
Sampling Systems™ GMP Pallet Knives
Product Code. 15899621 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
185 mm
225 mm
325 mm
380 mm
440 mm
495 mm
Length (Metric) Blade:
100 mm
120 mm
150 mm
200 mm
250 mm
300 mm
350 mm
Width (Metric) Blade:
20 mm
25 mm
32 mm
38 mm
45 mm
50 mm
Unit Size:
Each
Product Code. 15899621
Brand: Sampling Systems™ A330300
Only null left
Add to basket
Add to basket
Sampling Systems™ GMP Pallet Knives are a range of knives made from Stainless Steel that are ideal for hygienic applications.
  • Crevice free construction
  • All Stainless Steel construction
  • Satin finish
Specifications
Product Type Pallet Knife
Finish Polished
Length (Metric) Blade 300 mm
Width (Metric) Blade 45 mm
Material Stainless Steel
Length (Metric) 440 mm