missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GNE 9278

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GNE 9278 is a positive allosteric modulator of NMDA receptors. Potentiates NMDA receptors containing GluN2A, 2B, 2C and 2D subunits (EC50 values are 0.74, 3.07, 0.47 and 0.32 μM in a Ca2+ influx assay, respectively). Binds within the transmembrane domain and displays preference for agonist-bound NMDA receptors. Slows receptor deactivation and enhances the potency of glutamate and glycine .
Specifications
Specifications
| Quantity | 50 mg |
| Molecular Formula | C21H27N5O3S |
| Formula Weight | 429.54 |
| Product Type | Positive Allosteric Modulator |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction