missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GNF 5
GREENER_CHOICE

Product Code. 30002041
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002041

Brand: Tocris Bioscience™ 4908/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective allosteric inhibitor of Bcr-Abl; analog of GNF 2 (Cat. No. 4399)

Selective, non-ATP competitive allosteric inhibitor of Bcr-Abl (IC50 = 220 nM for wild-type Abl). Binds the myristate-binding site of Abl. Acts in combination with nilotinib to inhibit T315I Bcr-Abl in vitro and in vivo. Analog of GNF 2 (Cat. No. 4399).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778277-15-9
Molecular Formula C20H17F3N4O3

Specifications

Quantity 10 mg
Formula Weight 418.37
Molecular Weight (g/mol) 418.376
Chemical Name or Material GNF 5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.