missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72567
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
IgG (H+L) | |
Polyclonal | |
Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Alkaline Phosphatase (AP) | |
0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
Dog IgG whole molecule | |
1 mg | |
Adaptive Immunity, Immunology | |
Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction