missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
150.00€ - 299.00€
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Concentration | 4.5mg/mL |
| Dilution | Flow Cytometry 1:20-1:2000, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18272985
|
Novus Biologicals
NBP2-60652 |
1 mg |
299.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681489
|
Novus Biologicals
NBP2-60652-0.5mg |
0.5 mg |
150.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy.Specifications
| IgG (H+L) | |
| Flow Cytometry 1:20-1:2000, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000 | |
| Unconjugated | |
| Adaptive Immunity, Immunology | |
| IgG | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy gamma chains on dog IgG and light chains on all dog immunoglobulins |
| 4.5mg/mL | |
| Polyclonal | |
| Goat | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.05% Sodium Azide | |
| Secondary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title