missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Duck IgM Fc Secondary Antibody, Novus Biologicals™

Product Code. 18611029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18611029 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18611029 Supplier Novus Biologicals Supplier No. NBP269680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgM Fc Polyclonal antibody specifically detects IgM Fc in Duck samples. It is validated for Western Blot, ELISA, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM Fc
Applications Western Blot, ELISA, Dot Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Formulation Lyophilized from PBS (pH 7.2)
Host Species Goat
Immunogen Purified IgM isolated from duck serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Duck
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.